Please login to your online account to display your discounted pricing

SR 57227 hydrochloride, Tocris


Potent, selective 5-HT3 agonist

Quantity 10mg

Manufacturer: r & d systems  RDS120510

 View more versions of this product

Catalog No. 120510

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material SR 57227 hydrochloride
Quantity 10mg
CAS 77145-61-0
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C10H15Cl2N3·HCl
Formula Weight 248.15
Synonym 1-(6-Chloro-2-pyridinyl)-4-piperidinamine hydrochloride

Specifications

Chemical Name or Material SR 57227 hydrochloride
Assay Percent Range >99%
Molecular Formula C10H15Cl2N3·HCl
Formula Weight 248.15
Synonym 1-(6-Chloro-2-pyridinyl)-4-piperidinamine hydrochloride
Accessories

Store at Room Temperature