Please login to your online account to display your discounted pricing

SR 57227 hydrochloride, Tocris

Potent, selective 5-HT3 agonist

$164.61 - $625.50

Chemical Identifiers

Chemical Name or Material SR 57227 hydrochloride
Assay Percent Range >99%
Molecular Formula C10H15Cl2N3·HCl
Formula Weight 248.15
Synonym 1-(6-Chloro-2-pyridinyl)-4-piperidinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

120510

 
r & d systems
RDS120510
10mg Each for $164.61

120550

 
r & d systems
RDS120550
50mg Each for $625.50
Description & Specifications

Specifications

Formula Weight 248.15
Quantity 10mg
CAS 77145-61-0
Chemical Name or Material SR 57227 hydrochloride
Molecular Formula C10H15Cl2N3·HCl
Assay Percent Range >99%
Synonym 1-(6-Chloro-2-pyridinyl)-4-piperidinamine hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature