Please login to your online account to display your discounted pricing

SR 2640 hydrochloride, Tocris

Potent, selective LTD4 /LTE4 receptor antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material SR 2640 hydrochloride
Assay Percent Range >99%
Molecular Formula C23H18N2O3·HCl
Formula Weight 406.87
Synonym 2-[[3-(2-Quinolinylmethoxy)phenyl]amino]benzoic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

180410

 
r & d systems
RDS180410
10mg Each for $177.37

180450

 
r & d systems
RDS180450
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 406.87
Quantity 10mg
Chemical Name or Material SR 2640 hydrochloride
Molecular Formula C23H18N2O3·HCl
CAS 146662-42-2
Assay Percent Range >99%
Synonym 2-[[3-(2-Quinolinylmethoxy)phenyl]amino]benzoic acid hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature