missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SR 1664, Tocris™

High affinity PPARγ ligand; blocks Cdk5-dependent PPARγ phosphorylation

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >97%
Molecular Formula C33H29N3O5
Formula Weight 547.6
Synonym 4'-[[2, 3-Dimethyl-5-[[[(1S)-1-(4-nitrophenyl)ethyl]amino]carbonyl]-1H-indol-1-yl]methyl]-[1, 1'-biphenyl]-2-carboxylic acid
Chemical Name or Material SR 1664
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
440910 R & D Systems
440910
10mg Each for $244.81
440950 R & D Systems
440950
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>97%
547.6
SR 1664
C33H29N3O5
4'-[[2, 3-Dimethyl-5-[[[(1S)-1-(4-nitrophenyl)ethyl]amino]carbonyl]-1H-indol-1-yl]methyl]-[1, 1'-biphenyl]-2-carboxylic acid

Specifications

10mg
>97%
547.6
SR 1664
1338259-05-4
C33H29N3O5
4'-[[2, 3-Dimethyl-5-[[[(1S)-1-(4-nitrophenyl)ethyl]amino]carbonyl]-1H-indol-1-yl]methyl]-[1, 1'-biphenyl]-2-carboxylic acid

Safety and Handling

Recommended Storage : Store at -20°C