Please login to your online account to display your discounted pricing

SR 16584, Tocris™

Selective α3β4 nAChR antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SR 16584
Assay Percent Range >98%
Molecular Formula C17H22N2O
Formula Weight 270.37
Synonym 1, 3-Dihydro-1-(3-exo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indol-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

442410

 
r & d systems
442410
10mg Each for $252.39

442450

 
r & d systems
442450
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 270.37
Quantity 10mg
Synonym 1, 3-Dihydro-1-(3-exo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indol-2-one
Assay Percent Range >98%
Chemical Name or Material SR 16584
Molecular Formula C17H22N2O
CAS 1150153-86-8
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C