Please login to your online account to display your discounted pricing

SR 142948, Tocris

Highly potent NT receptor antagonist

$162.94 - $349.10

Chemical Identifiers

Chemical Name or Material SR 142948
Assay Percent Range >97%
Molecular Formula C39H51N5O6
Formula Weight 685.86
Synonym 2-[[[5-(2, 6-Dimethoxyphenyl)-1-[4-[[[3-(dimethylamino)propyl]methylamino]carbonyl]-2-(1-methylethyl)phenyl]-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13, 7]decane-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

23091

 
r & d systems
23091
1mg Each for $162.94

230910

 
r & d systems
230910
10mg Each for $349.10
Description & Specifications

Specifications

Formula Weight 685.86
Quantity 1mg
Chemical Name or Material SR 142948
Assay Percent Range >97%
CAS 184162-64-9
Synonym 2-[[[5-(2, 6-Dimethoxyphenyl)-1-[4-[[[3-(dimethylamino)propyl]methylamino]carbonyl]-2-(1-methylethyl)phenyl]-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13, 7]decane-2-carboxylic acid
Molecular Formula C39H51N5O6
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C