Please login to your online account to display your discounted pricing

SR 11237, Tocris™

Pan RXR agonist

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material SR 11237
Assay Percent Range >98%
Molecular Formula C24H28O4
Formula Weight 380.48
Synonym 4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1,3-dioxolan-2-yl]-benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

341110

 
r & d systems
RDS341110
10mg Each for $238.81

341150

 
r & d systems
RDS341150
50mg Each for $984.19
Description & Specifications

Specifications

Formula Weight 380.48
Quantity 10mg
Chemical Name or Material SR 11237
Assay Percent Range >98%
Molecular Formula C24H28O4
CAS 146670-40-8
Synonym 4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1,3-dioxolan-2-yl]-benzoic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C