Please login to your online account to display your discounted pricing

SR 1001, Tocris™

RORα/γ inverse agonist

$233.76 - $963.39

Chemical Identifiers

Chemical Name or Material SR 1001
Assay Percent Range >98%
Molecular Formula C15H13F6N3O4S2
Formula Weight 477.4
Synonym N-[4-Methyl-5-[[[4-[2, 2, 2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]amino]sulfonyl]-2-thiazolylacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

486810

 
r & d systems
RDS486810
10mg Each for $233.76

486850

 
r & d systems
RDS486850
50mg Each for $963.39
Description & Specifications

Specifications

Formula Weight 477.4
Quantity 10mg
Chemical Name or Material SR 1001
Assay Percent Range >98%
Molecular Formula C15H13F6N3O4S2
CAS 1335106-03-0
Recommended Storage Store at +4°C
Synonym N-[4-Methyl-5-[[[4-[2, 2, 2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]amino]sulfonyl]-2-thiazolylacetamide
Safety and Handling

Store at +4°C