Learn More
Squaric Acid Dibutyl Ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 133170010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Squaric acid dibutyl ester | |
Colorless to Yellow | |
121.0°C to 122.0°C (0.2 mmHg) | |
Authentic | |
Glass bottle | |
1.4930 to 1.4950 | |
1 g | |
15, 8897 | |
Solubility in water: insoluble. | |
CCCCOC1=C(OCCCC)C(=O)C1=O | |
226.27 | |
CHEBI:53612 | |
97% |
2892-62-8 | |
1.0500g/mL | |
>110°C | |
96% min. (GC) | |
C12H18O4 | |
MFCD00037150 | |
1.05 | |
dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p | |
XBRWELTXMQSEIN-UHFFFAOYSA-N | |
3,4-dibutoxycyclobut-3-ene-1,2-dione | |
65108 | |
226.27 | |
Liquid |
Chemical Identifiers
2892-62-8 | |
226.27 | |
XBRWELTXMQSEIN-UHFFFAOYSA-N | |
65108 | |
3,4-dibutoxycyclobut-3-ene-1,2-dione |
C12H18O4 | |
MFCD00037150 | |
dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p | |
CHEBI:53612 | |
CCCCOC1=C(OCCCC)C(=O)C1=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
GHS Signal Word: Warning
RUO – Research Use Only