Please login to your online account to display your discounted pricing

Alfa Aesar™ Squalene, 98%

$57.70 - $179.31

Chemical Identifiers

Chemical Name or Material Squalene
CAS 111-02-4
Molecular Formula C30H50
Formula Weight 410.73
MDL Number MFCD00008912
Merck Index 14,8768
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2094422

  SDS
alfa aesar
B2094422
100g Each for $57.70

AAB2094436

  SDS
alfa aesar™
B2094436
500g Each for $179.31
Description & Specifications

Specifications

Formula Weight 410.73
Quantity 100g
TSCA TSCA
Assay 98%
MDL Number MFCD00008912
Merck Index 14,8768
Recommended Storage 2° to 8°C
RTECS Number XB6010000
Density 0.858
Physical Form Liquid
Chemical Name or Material Squalene
Assay Percent Range 0.97
Molecular Formula C30H50
CAS 111-02-4

Applications
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

Solubility
Miscible with diethyl ether and acetone. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Safety and Handling

2° to 8°C