missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ SQ 22536, 95%

Manufacturer:  Alfa Aesar™ J67455MA

 View more versions of this product

Catalog No. AAJ67455MA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

95%
-30°C to -10°C
Solid
205.22
C9H11N5O
160° to 161°C
17318-31-9
9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine
White to off-white
10mg