missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals SQ 22536, 95%
Supplier: Thermo Scientific Chemicals J67455LB0
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5 mg | |
-30°C to -10°C | |
17318-31-9 | |
205.22 | |
160°C to 161°C | |
UKHMZCMKHPHFOT-UHFFFAOYNA-N | |
9-(oxolan-2-yl)-9H-purin-6-amine |
White to off-white | |
95% | |
C9H11N5O | |
9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine | |
Solid | |
NC1=C2N=CN(C3CCCO3)C2=NC=N1 | |
205.22 |
Chemical Identifiers
17318-31-9 | |
C9H11N5O | |
9-THF-Ade; 6-Amino-9-(tetrahydro-2-furyl)purine |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only