Please login to your online account to display your discounted pricing
American Educational Products Spring Binding Knife Switches
Durable components for classroom use.
Manufacturer: american educational products  P56015
 View more versions of this product
Catalog No. S78773
  • / Each
Description & Specifications
Specifications
Product Type Knife Switch
Material Bakelite™ Plastic
For Use With (Application) Bell or stranded wire to hook up electrical circuits without the need for banana cables
Includes Double pole, double throw with six spring terminals
For Use With (Equipment) Bell or stranded wire