American Educational Products Spring Binding Knife Switches
Durable components for classroom use.
Manufacturer:  American Educational Products P56014
 View more versions of this product
Catalog No. S78772
Specifications Description & Specifications
Specifications
Bell or stranded wire to hook up electrical circuits without the need for banana cables
Single pole, double throw with three spring terminals
Bakelite™ Plastic
Bell or stranded wire
Knife Switch