American Educational Products Spring Binding Knife Switches
Durable components for classroom use.
Manufacturer:  American Educational Products P56031
 View more versions of this product
Catalog No. S78771
Description
Description
  • Use with bell or stranded wire to hook up electrical circuits without the need for banana cables
  • Switches formed in a Bakelite™ plastic
  • Up to five wires can be held in place for the most elaborate circuits
Specifications
Specifications
Classroom use
Single pole, single throw with two spring terminals
Bakelite™ Plastic
Bell or stranded wire
Spring Binding Post Lamp Holders And Switch
Documents