Contec™ Sporicidin™ Disinfectant Wipes
Sporicidin™ wipes are EPA registered for use in hospitals, medical, dental fields, meets EPAâs criteria for use against SARS-CoV-2
Supplier: Contec™ CAN18012FCAN
Description
- Used in hospital, dental and medical environments since 1978
- Kills disease and odor-causing organisms
- Broad spectrum disinfectant- no rotation of intermediate disinfectants needed
- EPA registered in all 50 states and Puerto Rico
- OSHA bloodborne pathogen standards (29 CFR 1910.1030) compliant
- EPA toxicity rating low/very low
- Cleans, disinfects and deodorizes in one step
- Kills bacteriostatic activity up to 6 months, if not removed from surface
- Fungicidal, bactericidal, virucidal, mildewcidal and tuburculocidal
- Non-corrosive to surfaces including plastics, latex, vinyl, glass, treated wood, metal and glazed porcelain
- Contains pharmaceutical active ingredient system with no harsh chemicals
- Mildly alkaline (pH 8.5)
- Non-staining, non-flammable and non-abrasive to most surfaces
- Non-hazardous formula with no special shipping or disposal restrictions
- Alcohol and bleach-free wipes safe for medical and dental equipment
- Wipes ideal for office or lab use
- EPA Registration Number: 8383-7
Specifications
Disinfectant Wipes | |
For non-corrosive surfaces like plastics, latex, vinyl, glass, treated wood, metal and glazed porcelain surfaces in hospitals, dental and medical environments | |
180 Wipes/Pk., 12 Pk./Cs. |