Please login to your online account to display your discounted pricing

Sponge Nickel, A-5000, nonpromoted, Alfa Aesar™

Slurry in water

Chemical Identifiers

Chemical Name or Material Nickel
Name Note Nonpromoted
Physical Form Sponge A-5000
Molecular Formula Ni
MDL Number MFCD00167392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4486722

  SDS
alfa aesar
4486722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4486736

  SDS
alfa aesar
4486736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Color Silver
DOT Information DOT Class: 4.2, Packaging Group: II
Chemical Name or Material Nickel
Density 8.908g/cm3 at 20°C
Name Note Nonpromoted
Physical Form Sponge A-5000
Molecular Formula Ni
MDL Number MFCD00167392
Odor Odorless
Health Hazard 1 H251-H351-H317: Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction
Appearance Silver colored sponge
Safety and Handling

H251-H351-H317: Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction