missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sponge Nickel, A-5000, nonpromoted, Alfa Aesar™

Slurry in water

Manufacturer:  Alfa Aesar 4486722

 View more versions of this product

Catalog No. AA4486722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nickel
Nonpromoted
Silver colored sponge
HNO3
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
Odorless
944
Sponge A-5000
100g
7697-37-2
8.908g/cm3 at 20°C
MFCD00167392
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107
Silver

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00167392
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]
Safety and Handling

Safety and Handling

H251-H351-H317: Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.2, Packaging Group: II

SDS Documents