missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Spoked Ratchet Nut

For use with FastPrep-24

Manufacturer:  MP Biomedicals™ 116004510

Catalog No. 116004510



Description

Description

Securing knob for FastPreps-24™ used to lock the tube older on 3 step assembly.

Order Info

dimType

Specifications

Specifications

2Pack
Spoked Ratchet Nut
Documents