missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Splitomicin, Tocris

Sir2p inhibitor

$181.19 - $762.58

Chemical Identifiers

Assay Percent Range >97%
Molecular Formula C13H10O2
Formula Weight 198.22
Synonym 1, 2-Dihydro-3H-naphtho[2, 1-b]pyran-3-one
Chemical Name or Material Splitomicin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
154250 R & D Systems
RDS154250
50mg Each for $762.58
154210 R & D Systems
RDS154210
10mg Each for $181.19
Specifications Description & Specifications

Chemical Identifiers

>97%
198.22
Splitomicin
C13H10O2
1, 2-Dihydro-3H-naphtho[2, 1-b]pyran-3-one

Specifications

5690-03-9
C13H10O2
1, 2-Dihydro-3H-naphtho[2, 1-b]pyran-3-one
Splitomicin
>98%
198.22
50mg

Safety and Handling

Recommended Storage : Store at -20°C