Please login to your online account to display your discounted pricing

Spiro-(1,3-dioxolane-2,3'indolin)-2'-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Spiro-(1, 3-dioxolane-2, 3'indolin)-2'-one
CAS 6714-68-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO3
Formula Weight 191.19
MDL Number MFCD00173689
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S13949EA

 
fisher scientific company
S13949EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Spiro-(1, 3-dioxolane-2, 3'indolin)-2'-one
Assay Percent Range 97%
Molecular Formula C10H9NO3
Packaging Amber glass bottle
MDL Number MFCD00173689
CAS 6714-68-7
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.