Please login to your online account to display your discounted pricing

Spiro-(1,3-dioxolane-2,3'indolin)-2'-one, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  S13949EA

 View more versions of this product

Catalog No. S13949EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Chemical Name or Material Spiro-(1, 3-dioxolane-2, 3'indolin)-2'-one
Molecular Formula C10H9NO3
Assay Percent Range 97%
MDL Number MFCD00173689
CAS 6714-68-7
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Spiro-(1, 3-dioxolane-2, 3'indolin)-2'-one
CAS 6714-68-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO3
Formula Weight 191.19
MDL Number MFCD00173689
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.