Bel-Art™ SP Scienceware™ Spinwedge™ Magnetic Stirring Bars
For churning sediment or dissolving salts.
Manufacturer:  Bel-Art™ F371230003
Catalog No. 22-261724
Description
Description
  • Provides strong turbulence at low speeds
  • Wedge shape design plows up sediment for thorough dispersion
  • Sharply-angled broad face provides strong mixing action for viscous solutions
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclave safe
Specifications
Specifications
Yes
Provides strong turbulence at low speeds
0.5 in.
50.8mm
PTFE (Polytetrafluoroethylene)
2 in.
Durable and inert
Durable and inert
12.7mm
Wedge
Documents