Bel-Art™ SP Scienceware™ SpinPak Octagon Magnetic Stirring Bar Packs
Interrupted profile provides greater surface area and added turbulence compared to the smooth surface of cylindrical bars.
Manufacturer: Bel-Art™ F371570015
Description
- With integral pivot ring to reduce friction and chattering
- One size per pack
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
- Durable and inert
- Autoclavable
Specifications
Yes | |
Alnico magnet | |
8mm | |
6 | |
Stirring Bar Pack |
0.312 in. | |
1.5 in. | |
38mm | |
Octagonal |