Please login to your online account to display your discounted pricing

Spinel Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™

100 Orientation

$173.93 - $640.34

Chemical Identifiers

Chemical Name or Material Spinel Substrate
Name Note Polished One Side
Physical Form Plate
Molecular Formula MgAl2O4
MDL Number MFCD00049475
Synonym Magnesium aluminum oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38505KT

  SDS
alfa aesar
38505KT
1 ea. Each for $173.93

AA38505KU

  SDS
alfa aesar
38505KU
5 ea. Each for $640.34
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Spinel Substrate
Name Note Polished One Side
Physical Form Plate
Molecular Formula MgAl2O4
MDL Number MFCD00049475
Synonym Magnesium aluminum oxide