missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spinal Diluting Fluid (Crystal Violet), Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,64-19-7,548-62-9
CAS Min % 90.0,7.88,0.1
CAS Max % 94.0,8.2,0.11
Chemical Name or Material SPINAL DILUTING FLUID Crystal Violet- Acetic Acid Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
7996-16 Ricca Chemical Company
RI799616
Poly Bottle 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
7996-4 Ricca Chemical Company
RI79964
120mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,64-19-7,548-62-9
90.0,7.88,0.1
94.0,8.2,0.11
SPINAL DILUTING FLUID Crystal Violet- Acetic Acid Solution

Specifications

500mL
94.0,0.11,8.2
0.1,90.0,7.88
7732-18-5,64-19-7,548-62-9
Poly Bottle
SPINAL DILUTING FLUID Crystal Violet- Acetic Acid Solution