Liquid Safety Solutions Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
Manufacturer:  Liquid Safety Solutions 450020
 View more versions of this product
Catalog No. 19-122-867
Description
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
  • Contain polyproylene for superior absorption, plus polyester casing for excellent stretching ability
  • Surround spill with socks to prevent spreading or to use as a barricade
  • Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
Pillows
  • Pillows soak up different size spills in ponds, ditches, open water or under machinery
  • Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
  • Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
Drum Tops
  • Precut hole provides a perfect fit around pump
  • Superior absorbency keeps drum tops clean
  • Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Soaker Roll/Pad
  • Provides absorption protection from spills and cushioning to minimize breakage
  • Sorbent Pad
    • Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
  • Spill Kit
    • Complete spill response capability for either inside or outside storage
    Specifications
    Specifications
    Complete spill response capability for either inside or outside storage
    1 Kit
    64.26 L
    28 lb.
    Convenient lever-lock metal ring tops, five 3 in. x 4 ft. socks, hundred 16 x 18 in. pads, one disposal bag and twist tie, one pair nitrile gloves, one pair splash goggles, one 20 gal. poly drum
    17 gal.
    Universal Sorbent Spill Kit
    12.7 kg