Liquid Safety Solutions Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
Manufacturer:  Liquid Safety Solutions DB75
 View more versions of this product
Catalog No. 19-066-562
Description
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
  • Contain polyproylene for superior absorption, plus polyester casing for excellent stretching ability
  • Surround spill with socks to prevent spreading or to use as a barricade
  • Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
Pillows
  • Pillows soak up different size spills in ponds, ditches, open water or under machinery
  • Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
  • Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
Drum Tops
  • Precut hole provides a perfect fit around pump
  • Superior absorbency keeps drum tops clean
  • Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Soaker Roll/Pad
  • Provides absorption protection from spills and cushioning to minimize breakage
  • Sorbent Pad
    • Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
  • Spill Kit
    • Complete spill response capability for either inside or outside storage
    Specifications
    Specifications
    Gray
    Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
    18 L x 16 in. W (45.72 x 40.64 cm)
    120.96 L
    45.72 cm
    16 lb.
    16 in.
    18 x 16 in. (45.72 x 40.64 cm)
    100/Cs.
    32 gal.
    18 in.
    Universal Sorbent Pad
    7.25 kg
    40.64 cm