Liquid Safety Solutions Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
Manufacturer:  Liquid Safety Solutions G37*
 View more versions of this product
Catalog No. 19-066-572
Description
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
  • Contain polyproylene for superior absorption, plus polyester casing for excellent stretching ability
  • Surround spill with socks to prevent spreading or to use as a barricade
  • Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
Pillows
  • Pillows soak up different size spills in ponds, ditches, open water or under machinery
  • Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
  • Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
Drum Tops
  • Precut hole provides a perfect fit around pump
  • Superior absorbency keeps drum tops clean
  • Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Soaker Roll/Pad
  • Provides absorption protection from spills and cushioning to minimize breakage
  • Sorbent Pad
    • Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
  • Spill Kit
    • Complete spill response capability for either inside or outside storage
    Specifications
    Specifications
    Gray
    Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
    6/Cs.
    17 gal.
    10 ft.
    Polyester, Polypropylene
    14 lb.
    3 in.
    10 ft. x 3 in. (3 m x 7.62 cm)
    Spills, fine fiber Sustayn&trade, sorbent
    10 ft. L x 3 in. W (3 m x 7.62 cm)
    64.26 L
    301 cm
    Universal Sorbent Sock
    6.35 kg
    7.62 cm