NPS Corp. Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
Manufacturer: NPS Corp. G34*
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
- Contain polyproylene for superior absorption, plus polyester casing for excellent stretching ability
- Surround spill with socks to prevent spreading or to use as a barricade
- Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
- Pillows soak up different size spills in ponds, ditches, open water or under machinery
- Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
- Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
- Precut hole provides a perfect fit around pump
- Superior absorbency keeps drum tops clean
- Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
- Provides absorption protection from spills and cushioning to minimize breakage
- Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
- Complete spill response capability for either inside or outside storage
Specifications
44 gal. | |
44 gal., 166.32L | |
Gray | |
Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy | |
4 ft. | |
40 Case | |
14.06kg | |
7.62cm |
166.32L | |
Polyester | |
4 ft. x 3 in. (121 x 7.62cm) | |
Spills, fine fiber Sustayn™ sorbent | |
121cm | |
31 lb. | |
3 in. | |
Universal Sorbent Sock |