NPS Corp. Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
Manufacturer:  NPS Corp. G111
 View more versions of this product
Catalog No. 19-066-569
Description
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
  • Contain polyproylene for superior absorption, plus polyester casing for excellent stretching ability
  • Surround spill with socks to prevent spreading or to use as a barricade
  • Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
Pillows
  • Pillows soak up different size spills in ponds, ditches, open water or under machinery
  • Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
  • Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
Drum Tops
  • Precut hole provides a perfect fit around pump
  • Superior absorbency keeps drum tops clean
  • Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Soaker Roll/Pad
  • Provides absorption protection from spills and cushioning to minimize breakage
  • Sorbent Pad
    • Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
  • Spill Kit
    • Complete spill response capability for either inside or outside storage
    Specifications
    Specifications
    11 gal.
    11 gal., 41.58L
    10 x 10 in. (25.4 x 25.4cm)
    Low profile Plastic pan, spills, fine fiber sorbent
    25.4cm
    15 lb.
    10 in.
    Universal Sorbent Pillow Kit
    41.58L
    Polyester
    Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
    10 in.
    12 Case
    6.8kg
    25.4cm