Liquid Safety Solutions Spilfyter™ Universal Pads and Rolls
Universal sorbents for acids or bases, and nonaggressive spills, such as oil, coolants, solvents, and water
Manufacturer:  Liquid Safety Solutions DB95
 View more versions of this product
Catalog No. 19-066-565
Description
Description
Sorbents are manufactured with eight layers of high-quality, fine fiber polyethylene. Premium sorbents are bonded every 4 in., Economy every 8 in.
  • For better fit in irregular spaces, rolls are perforated horizontally every 12 in.and vertically down the center of the roll
  • For more flexible fit in narrow spaces, pads are perforated vertically
  • Premium pads and rolls offer extended use and high durabilty
  • Economy pads and rolls are ideal for one-time applications
  • Use pads around machinery, to line shelves or for cleanups
  • Use rolls in assembly and production areas
  • Denim Blue color inconspicuously hides dirt and grime for MRO applications
Specifications
Specifications
OSHA 29 CFR 1910
300 ft. x 32 in. (91.4 m x 81.3 cm)
Single-weight
242.2 L
91.4 m
Universal Sorbent Roll
15 kg
81.3 cm
Denim Blue
Acids or bases, and non-aggressive spills, as oil, coolants, solvents and water
64 gal.
300 ft.
Polyethylene Fiber
32 lb.
32 in.