Please login to your online account to display your discounted pricing

Spilfyter™ Oil-Only Containment Products: Pillows

Use components together for optimum performance

$129.59 - $130.53

Specifications

Certifications/Compliance NFPA
Color Blue
Material Polyester Casing, Polypropylene Fill
For Use With (Application) For standard use applications
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Weight (English) Product Type Price Quantity    

19-142-782

 
national packaging
M62*
18 x 18 in. (45.72 x 45.72cm) 11 lb. Oil Only Pillow Case of 10 Each for $129.59

19-066-597

 
national packaging
M61*
10 x 10 in. (25.4 x 25.4cm) 8 lb. Oil Only Sorbent MRO Pillow Case of 20 Each for $130.53
Description & Specifications

Specifications

Certifications/Compliance NFPA
Color Blue
Material Polyester Casing, Polypropylene Fill
For Use With (Application) For standard use applications

Fine fibers and state-of-the-art multilayering technology provide superior capillary action.

Pillows

  • Pillows soak up spills in ponds, ditches, open water or under machinery

Static-resistance sorbents meet NFPA specifications.