missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spilfyter™ Oil-Only Containment Products: Booms

Use components together for optimum performance

$130.51 - $238.76

Specifications

Certifications/Compliance NFPA
Color White
Material Meltblown Fiber
For Use With (Application) Used in oil, gas, mining or fracking industry for spill control response or prevention
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Dimensions (L x W) Weight (English) Product Type Price Quantity  
Catalog Number Mfr. No. Capacity Dimensions (L x W) Weight (English) Product Type Price Quantity  
19-066-595 National Packaging
M55
38 gal., 143L 10 ft. x 5 in. (3m x 12.7cm) 18 lb. Oil Only Sorbent Premium Boom Case of 4 Each for $148.34
19-066-593 National Packaging
M40*
75 gal., 283L 20 ft. x 8 in. (6m x 20.32cm) 40 lb. Oil Only Sorbent Premium Boom Case of 2 Each for $238.76
19-066-596 National Packaging
M58
10 gal., 35L 16 ft. x 8 in. (4.87m x 20.32cm) 7 lb. Oil Only Sorbent Bilge Boom Case of 10 Each for $130.51
19-066-594 National Packaging
JVM50
75 gal., 283L 10 ft. x 8 in. (3m x 20.32cm) 40 lb. Oil Only Sorbent Premium Boom Case of 4 Each for $159.17
Specifications

Description

Fine fibers and state-of-the-art multilayering technology provide superior capillary action.

Compliance

Static-resistance sorbents meet NFPA specifications.

Description & Specifications

Specifications

NFPA
White
Meltblown Fiber
Used in oil, gas, mining or fracking industry for spill control response or prevention