Liquid Safety Solutions Spilfyter™ Decontamination Solutions
For washing unknown contaminants
Manufacturer:  Liquid Safety Solutions JV680045
 View more versions of this product
Catalog No. 19-066543
Description
Description
  • Wash solutions for cleaning contaminant spills
  • Ready-to-use, requiring no dilution
  • Phosphate-free
  • Available in five formulas
  • Delivered in 5 gallon carboys
Decon #1
  • For contaminants such as inorganic acids, metal processing wastes, solvents and organic materials, PBBs, PCBs, and Etiological materials
  • Decon #2
    • For heavy metals (Mercury, Lead, Cadmium, etc.), pesticides, chlorinated phenols, dioxins and non-acid inorganic wastes
    • Expiration date of 1 year
    Decon #3
    • For solvents and inorganic compounds, PBBs, PCBs, oils and greases, or unspecified wastes not suspected to be contaminated with pesticides
    • Can also be used as a general purpose rinse for Decon Solutions 1 and 2
    Decon #5
    • Use as a final wash down
    Specifications
    Specifications
    OSHA 1910.120 1910.120(k)(2)(i)
    For use as final wash down; Decon solution
    1 Ea.
    18.92 L
    Decon Solution 5
    21 kg
    Green
    Liquid
    5 gal.
    Proprietary Ingredient, Water
    47 lb.