NPS Corp. Spilfyter™ Chemical Classifiers Test Strips and Charts
Test strips allow immediate color-coded identification of spilled chemicals
Manufacturer:  NPS Corp. JV570050
 View more versions of this product
Catalog No. 19-057467
Description
Description
Chemical Classifier Strips show responder if an unknown spill contains critical levels of these contaminant categories: acid/base, oxidizer, fluoride, petroleum/organic solvent, iodine/bromine/chlorine. Classifier Chart compares sample colors to standard color.
Use strips to identify unknowns in sewers, construction sites, accident sites, storage facilities, landfills, waste management facilities.
Order Info
Additional test strips and classifier charts are available separately.
Specifications
Specifications
Quickly assist the responder in classifying the chemical and wastewater products for spill proper treatment
20°C
1 lb.
50 Case
Chemical Classifier Test Strip
0.4kg