Please login to your online account to display your discounted pricing

Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  SPIKE1

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. SPIKE-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125mL
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr Tartaric Acid/tr HF:
Al    •200μg/mL
Sb    •50μg/mL
As    •200μg/mL
Ba    •200μg/mL
Be    •5μg/mL
Cd    •5μg/mL
Cr    •20μg/mL
Co    •50μg/mL
Cu    •25μg/mL
Fe    •100μg/mL
Pb    •50μg/mL
Mn    •50μg/mL
Ni    •50μg/mL
Se    •200μg/mL
Ag    •5μg/mL
Tl    •200μg/mL
V    •50μg/mL
Zn    •50μg/mL
Chemical Name or Material ICP Spike Sample 1
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Health Hazard 1 Causes skin irritation, serious eye irritation
Packaging Triple-leached LDPE bottle
Solvent or Matrix 5% HNO3/tr Tart. Acid/tr HF
DOT Information Hazardous
Purity Grade For CLP (Contract Laboratory Program)

Specifications

Chemical Name or Material ICP Spike Sample 1
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al200μg/mL
As200μg/mL
Ba200μg/mL
Se200μg/mL
Tl200μg/mL
Fe100μg/mL
Co50μg/mL
Mn50μg/mL
Ni50μg/mL
Pb50μg/mL
Sb50μg/mL
V50μg/mL
Zn50μg/mL
Cu25μg/mL
Cr20μg/mL
Ag5μg/mL
Be5μg/mL
Cd5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation

Ambient