Invitrogen™ SPHK2 Recombinant Human Protein DFS Item
SPHK2, along with SPHK1, catalyzes the phosphorylation of sphingosine producing the signalling molecule sphingosine-1-phosphate (S1P).
Manufacturer:  Invitrogen™ LSPV5217
 View more versions of this product
Catalog No. PV5217
Description
Description
S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapuetics for the treatment of cancer.
Specifications
Specifications
NP_064511.2
Frozen
96.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SPHK and DGCK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
SPHK2
Baculovirus
Human
GST Tag
Documents