SPEX™ SamplePrep Geno/Grinder™ Accessory, Grinding Mill
Compatible with all SPEX SamplePrep Titer Plates and Vial Sets
$558.81 - $558.81
Specifications
For Use With (Equipment) Titer plates and vial sets of Geno/Grinder
Material Silica
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Particle Size Price Quantity  
Catalog Number Mfr. No. Particle Size Price Quantity  
2166 SPEX™ SamplePrep
SPX2166
400 to 600μm Each for $558.81
Specifications
Description
  • Acid washed silica grinding beads
  • Bead size ranges from 800-1000μm
  • Suitable for basic cell lysis applications using the 2010 Geno/Grinder
  • Treated to remove fine particles and contaminates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Titer plates and vial sets of Geno/Grinder
Silica