SPEX™ SamplePrep Disposable XRF X-Cell™ Sample Cups Window Film
Strong and durable window film. SPEX SamplePrep Disposable XRF X-Cell Sample Cups Window Film covers the cell and serves as the sample window, allowing uniform transmission of X-rays.
Manufacturer:  SPEX™ SamplePrep 3526
 View more versions of this product
Catalog No. 3526
Description
Description
  • Minimizes leaks that could damage the spectrophotometer
Order Info
Available in a variety of materials, each resistant to a range of chemicals. All sizes are compatible with X-Cell sample cups.
Specifications
Specifications
Ultralene
6.35 cm
Window Film
0.16 mil
2.5 in.
500/box
2.5 in. dia. circles