SPEX™ SamplePrep Disposable XRF X-Cell™ Sample Cups Window Film
Strong and durable window film. SPEX SamplePrep Disposable XRF X-Cell Sample Cups Window Film covers the cell and serves as the sample window, allowing uniform transmission of X-rays.
$356.66 - $851.16
Specifications
Type Window Film
View More Specs
Products 5
Catalog Number Mfr. No. Quantity Material Thickness Price Quantity  
Catalog Number Mfr. No. Quantity Material Thickness Price Quantity  
04-675-1
View Documents Promotion Details
SPEX™ SamplePrep
SPX3511
1 Roll Kapton 0.33 mil
Each for $665.50
04-675-4
View Documents Promotion Details
SPEX™ SamplePrep
3516
1 Roll Mylar 0.12 mil
Each for $463.63
04-675-5
View Documents Promotion Details
SPEX™ SamplePrep
3517
1 Roll Mylar 0.25 mil
Each for $356.66
04-675-7
View Documents Promotion Details
SPEX™ SamplePrep
SPX3520
1 Roll Polypropylene 0.2 mil
Each for $567.16
3525
View Documents Promotion Details
SPEX™ SamplePrep
SPX3525
1 Roll Ultralene 0.16 mil
Each for $851.16
Description
Description
  • Minimizes leaks that could damage the spectrophotometer
Order Info
Available in a variety of materials, each resistant to a range of chemicals. All sizes are compatible with X-Cell sample cups.
Specifications
Specifications
Window Film
SDS
missing translation for 'documents'
missing translation for 'documents'