Cole-Parmer™ XRF Spec-Caps™ Cups
These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
Supplier: Cole-Parmer™ 3615
Description
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
- Reinforces sample disks to resist chipping and breaking.
- Makes sample disks easier to remove from the pellet die.
- Minimizes contamination during sample handling.
- Protects the bore of the pellet die from abrasive samples.
- Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Specifications
XRF Spectrometers | |
Produces pellets 35 mm x 5 mm | |
33 mm | |
8 mm | |
100 Pk. |
Sample Cup | |
1.2 in. | |
0.31 in. | |
Aluminum |