SPEX™ SamplePrep 6870 Freezer/Mill™
Large cryogenic impact grinder with self-contained liquid nitrogen tub and insulated case
Manufacturer:  SPEX™ SamplePrep 6870115
Catalog No. 68-701-15
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Touch-screen controls for programming grinding cycles, times, impact rate, pre-cooling and cooling times
  • Stores 10 grinding programs
  • Grinds samples in the 0.1 to 100g range
  • Holds one Large Vial, one Mid-Size Vial, or up to four Standard Small Vials
  • Auto-Fill System (6820) available for automated flling with liquid nitrogen
  • Safety features include LN sensor and lid interlock
  • Requires liquid nitrogen for operation
  • Supplied with choice of 6870L Accessory Package for Large Vials or 6870S Accessory Package for Small Vials
Specifications
Specifications
CE
Ideal for DNA or RNA extractions, materials testing, pharmaceutical analysis, drug testing, RoHS or WEEE and volatiles extraction
Self contained liquid Nitrogen tub and insulated case, 6820 auto fill system
3-prong grounded plug
0.0002 to 0.22 lb.
42 lb.
60Hz
115V 60Hz
One large vial, one mid size vial or up to four standard small vials
Touchscreen
Plant and animal tissue, seeds, Plastic and Polymers, Yeast, pharmaceuticals, food products, electronic components, textiles, hair, teeth, and bones
0.1 to 100g
19kg
115V