SPEX™ SamplePrep 6870 Freezer/Mill™ Grinding Vial Set
$1,280.33 - $1,618.00
Specifications
For Use With (Equipment) 6870 Freezer/Mill
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Capacity (English) Capacity (Metric) Price Quantity  
6871 SPEX™ SamplePrep
SPX6871
For grinding electronic components 0.01 to 0.11 lb. 5 to 50g Each for $1,618.00
68-85 SPEX™ SamplePrep
SPX6885
For grinding soft tissues without metal contamination 0.006 to 0.05 lb. 3 to 25g Each for $1,280.33
Specifications Description & Specifications
Specifications
6870 Freezer/Mill