Please login to your online account to display your discounted pricing
SPEX™ SamplePrep 6870 Freezer/Mill™ End Plugs
$430.87 - $430.87
Specifications
For Use With (Equipment) 6885 Vial Set
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Material Size Price Quantity    
68-85E
spex™ sampleprep
SPX6885E
For grinding soft plant and animal tissues without metal contamination Polycarbonate Pair for $430.87
Description & Specifications
Specifications
For Use With (Equipment) 6885 Vial Set