Please login to your online account to display your discounted pricing
SPEX™ SamplePrep 6770 Freezer/Mill™ Accessories: Chromium-free Vial Set
For use with Spex SamplePrep Freezer/Mill Cryogenic Grinder
$715.46 - $715.46
Specifications
For Use With (Application) RoHS/WEEE testing
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
6771
spex™ sampleprep
SPX6771
Cr-Free Grinding Vial Set Each for $715.46
Description & Specifications
Specifications
For Use With (Application) RoHS/WEEE testing
Chromium-free Vial Set 6771
  • Sample capacity: 0.5 to 4.0mL per vial
  • Includes chromium-free impactor, four polycarbonate center cylinders and two end plugs
  • Transparent plastic allows visual check of grinding process
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.