Please login to your online account to display your discounted pricing

SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards


SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the
QuEChERS analytical techniques (GC, GC/MS, LC/MS)

Manufacturer: spex certiprep™  SPXQUECRAOACTTP

 View more versions of this product

Catalog No. QUECRAOACTT

  • / Each
 

Description & Specifications

Specifications

Product Type QuEChERS standard- triphenylphosphate
For Use With (Application) For QuEChERS analytical techniques (GC, GC/MS, LC/MS)
Concentration 2μg/mL
Solvents Acetonitrile
Standard Concentration 2μg/mL
Standard Components Triphenylphosphate
Volume (Metric) 5mL