missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards
SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the
QuEChERS analytical techniques (GC, GC/MS, LC/MS)
$141.97 - $307.60
Specifications
For Use With (Application) | For QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) |
---|---|
Solvents | Acetonitrile |
Volume (Metric) | 5 mL |
Catalog Number | Mfr. No. | Concentration | Product Type | Standard Components | Standard Concentration | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Product Type | Standard Components | Standard Concentration | Price | Quantity | ||
QUECRAOACTT |
SPEX CertiPrep™
QUECRAOACTTP |
2 µg/mL | QuEChERS standard- triphenylphosphate | Triphenylphosphate | 2 µg/mL |
Each for $288.69
|
|
QUECRPCB138 |
SPEX CertiPrep™
QUECRPCB138 |
50 µg/mL | QuEChERS standard- PCB 138 | PCB 138 | 50 µg/mL |
Each for $141.97
|
|
QUECRPCB153 |
SPEX CertiPrep™
QUECRPCB153 |
50 µg/mL | QuEChERS standard- PCB 153 | PCB 153 | 50 µg/mL |
Each for $274.49
|
|
QUECR3765 |
SPEX CertiPrep™
QUECR3765 |
20 µg/mL | QuEChERS standard- triphenylphosphate | Triphenylphosphate | 20 µg/mL |
Each for $307.60
|
|
QUECRPCB52 |
SPEX CertiPrep™
QUECRPCB52 |
50 µg/mL | QuEChERS standard- PCB 52 | PCB 52 | 50 µg/mL |
Each for $274.49
|
|
QUECR4381 |
SPEX CertiPrep™
QUECR4381 |
50 µg/mL | QuEChERS standard- tris-(dichloroisopropyl)phosphate | Tris-(dichloroisopropyl)phosphate | 50 µg/mL |
Each for $293.40
|
|
Description
Single Component Reference Standards
- Volume: 5mL
Specifications
For QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) | |
Acetonitrile | |
5 mL |