missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards

SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the
QuEChERS analytical techniques (GC, GC/MS, LC/MS)

Manufacturer:  SPEX CertiPrep™ SPXQUECR4381

 View more versions of this product

Catalog No. QUECR4381



Description

Description

Single Component Reference Standards

  • Volume: 5mL

Specifications

Specifications

50 µg/mL
QuEChERS standard- tris-(dichloroisopropyl)phosphate
Tris-(dichloroisopropyl)phosphate
5 mL
For QuEChERS Analytical Techniques (GC, GC/MS, LC/MS)
Acetonitrile
50 µg/mL
Documents