missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SPEX CertiPrep™ SPEXQuE™ GC/MS QC Standards

Sensible solutions for pesticide extraction

$273.60 - $278.40

Specifications

For Use With (Application) Sensible Solutions for Pesticide Extraction
Solvents Acetonitrile
Volume (Metric) 5 mL
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Standard Components Standard Concentration Price Quantity  
Catalog Number Mfr. No. Product Type Standard Components Standard Concentration Price Quantity  
QUECRMSQC2 View Documents SPEX CertiPrep™
SPXQUECRMSQC2
GC/MS QC standard 2 Anthracene 100 µg/mL Each for $278.40
 
QUECRMSQC1 View Documents SPEX CertiPrep™
SPXQUECRMSQC1
GC/MS QC standard 1 PCB 138, PCB 153 50 µg/mL Each for $273.60
 
Description

Description

  • Solvent: Acetonitrile
  • Volume: 5mL

Specifications

Specifications

Sensible Solutions for Pesticide Extraction
Acetonitrile
5 mL